4-(Trifluoromethyl)thiobenzamide
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RF03219.1 RF03219.10 RF03219.50
MAYBRF03219.1EA 0 EUR
MAYBRF03219.1 MAYBRF03219.10 MAYBRF03219.50
4-(Trifluoromethyl)thiobenzamide
4-(Trifluoromethyl)thiobenzamide
Formule: C₈H₆F₃NS
Kookpunt: 254 °C (1013 hPa)
Smeltpunt: 135 °C
MDL Nummer: MFCD00051806
CAS nummer: 72505-21-6
UN: 2811
ADR: 6.1,III