Genistein ≥98.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G0272-1G
G0272-100MG
TCIAG0272-1GEA
44
EUR
TCIAG0272-1G
TCIAG0272-100MG
Genistein ≥98.0% (by HPLC)
Genistein
Formule:
C₁₅H₁₀O₅ Smeltpunt: 297…298 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00016952 CAS nummer: 446-72-0 EINECS: 207-174-9 Merck Index: 14,04391 |
|
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...