4-Aminobutyric acid
Leverancier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MOLE32215466-100GEA
0
EUR
MOLE32215466-100G
4-Aminobutyric acid
4-Aminobutyric acid
Formule:
C₄H₉NO₂ MW: 103,12 g/mol Kookpunt: 248 °C (1013 hPa) Smeltpunt: 202 °C Dichtheid: 1,21 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008226 CAS nummer: 56-12-2 EINECS: 200-258-6 Merck Index: 13,00429 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...