4-Butylbenzenesulphonyl chloride

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 4-Butylbenzene-1-sulphonyl chloride

BTB10243.1 BTB10243EA BTB10243.50
MAYBBTB10243.1EA 0 EUR
MAYBBTB10243.1 MAYBBTB10243EA MAYBBTB10243.50
4-Butylbenzenesulphonyl chloride
4-Butylbenzenesulphonyl chloride
Formule: C₁₀H₁₃ClO₂S
MDL Nummer: MFCD00173908
CAS nummer: 54997-92-1
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR