4-Chloro-3-nitrobenzenesulphonyl chloride
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
4-Chloro-3-nitrobenzenesulphonyl Chloride, 4-Chloro-3-nitrobenzene-1-sulphonyl chloride
KM08399.1
KM08399.10
KM08399.50
MAYBKM08399.1EA
0
EUR
MAYBKM08399.1
MAYBKM08399.10
MAYBKM08399.50
4-Chloro-3-nitrobenzenesulphonyl chloride
4-Chloro-3-nitrobenzenesulphonyl chloride
Formule:
C₆H₃Cl₂NO₄S MW: 256.07 g/mol Smeltpunt: 61.1 |
MDL Nummer:
MFCD00007440 CAS nummer: 97-08-5 EINECS: 202-558-2 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...