Butyltin hydroxide oxide, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: N-butyltinhydroxideoxide, Butylhydroxyoxostannane
M-Clarity™ quality level = MQ200
8.40126.0010 8.40126.0100
8.40126.0010EA 40.2 EUR
8.40126.0010 8.40126.0100
Butyltin hydroxide oxide, Sigma-Aldrich®
Butyltin hydroxide oxide
Formule: C₄H₁₀O₂Sn
MW: 208.83 g/mol
MDL Nummer: MFCD00013927
CAS nummer: 2273-43-0
EINECS: 218-880-1
UN: 3146
ADR: 6.1 III
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Molar Mass 208.81 g/mol
Boiling Point 350 °C
Vapor Pressure 1.47 hPa (20 °C)
Synonyms Butylhydroxyoxo stannane
Hill Formula C4H10O2Sn