Ethyl diethoxyacetate ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

L00390.14 L00390.22
L00390.14EA 46.8 EUR
L00390.14 L00390.22
Ethyl diethoxyacetate ≥97%
Ethyl diethoxyacetate

Purity: 97+% CAS No.: 6065-82-3 Molecular Formula: C8H16O4 Formula Weight: 176.21 Flash Point: 85°(185°F) Refractive Index: 1.4100 Boiling Point: 198-200° Beilstein Reference: 878579 MDL No.: MFCD00009231

Formule: C₈H₁₆O₄
Kookpunt: 198…200 °C
Dichtheid: 0,988
Vlampunt: 85 °C (185 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009231
CAS nummer: 6065-82-3
EINECS: 227-999-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR