Ethyl methoxyacetate ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Methoxyacetic ethyl ester
A18770.09
A18770.18
A18770.09EA
43.4
EUR
A18770.09
A18770.18
Ethyl methoxyacetate ≥98%
Ethyl methoxyacetate
Purity: 98%
CAS No.: 3938-96-3
Molecular Formula: CH3OCH2CO2CH2CH3
Formula Weight: 118.13
Flash Point: 46°(115°F)
Refractive Index: 1.401
Boiling Point: 44-46°/9mm
Beilstein Reference: 1744761
UN No.: 3272
MDL No.: MFCD00009176
Formule:
C₅H₁₀O₃ MW: 118.13 g/mol Kookpunt: 44…46 °C (9 mmHg) Dichtheid: 1.00 Vlampunt: 46 °C (115 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00009176 CAS nummer: 3938-96-3 EINECS: 223-521-7 UN: 3272 ADR: 3 III |