Acrylzuur, low water content ≥99.5% stabilised

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Prop-2-enoic acid

Stabilisatie: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)

43359.K4 43359.AP
43359.K4EA 164 EUR
43359.K4 43359.AP
Acrylzuur, low water content ≥99.5% stabilised
Acrylzuur

Stabilized with ca 200 ppm 4-methoxyphenol


Purity: 99.5% CAS No.: 79-10-7 Molecular Formula: H2C=CHCO2H Formula Weight: 72.06 Flash Point: 54°(130°F) Physical Form: Liquid Density: 1.051 Refractive Index: 1.4202 Odour: Pungent Melting Point: ca 13° Boiling Point: 138-139° Solubility: Miscible with water, alcohols, ether Beilstein Reference: 635743 Merck Reference: 14,130 UN No.: 2218 MDL No.: MFCD00004367

Formule: C₃H₄O₂
Kookpunt: 138…139 °C
Smeltpunt: ∼ 13 °C
Dichtheid: 1,051
Vlampunt: 54 °C (130 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004367
CAS nummer: 79-10-7
EINECS: 201-177-9
UN: 2218
ADR: 8,II
Merck Index: 14,130

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR