Thiobenzamide 95%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
164730100 164730500
ACRO164730100EA 26 EUR
ACRO164730100 ACRO164730500
Thiobenzamide 95%
Thiobenzamide
Formule: C₇H₇NS
Kookpunt: 245 °C (1013 hPa)
Smeltpunt: 114…118 °C
MDL Nummer: MFCD00008060
CAS nummer: 2227-79-4
EINECS: 218-765-6
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Yellow-green Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 114 to 118 °C
HPLC ≥94 %