Cesium trifluoroacetate
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
44633.09
44633.18
44633.09EA
45.9
EUR
44633.09
44633.18
Cesium trifluoroacetate
Cesium trifluoroacetate
CAS No.: 21907-50-6
Molecular Formula: CsOOCCF3
Formula Weight: 245.92
Physical Form: Crystalline
Melting Point: 115°
MDL No.: MFCD00051304
Formule:
C₂CsF₃O₂ Smeltpunt: 115 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00051304 CAS nummer: 21907-50-6 |