Ethyl trifluoroacetate

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SB01125.1 SB01125.10 SB01125.50
MAYBSB01125.1EA 0 EUR
MAYBSB01125.1 MAYBSB01125.10 MAYBSB01125.50
Ethyl trifluoroacetate
Ethyl trifluoroacetate
Formule: C₄H₅F₃O₂
Kookpunt: 61 °C (1013 hPa)
Smeltpunt: –78 °C
Dichtheid: 1,19 g/cm³ (20 °C)
Vlampunt: –17 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000419
CAS nummer: 383-63-1
EINECS: 206-851-6
UN: 2924
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR