Afsluitkranen, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6460-0002 6460-0004
229-0016EA 55.9 EUR
229-0016 229-0017
Afsluitkranen, Nalgene®
Afsluitkranen
Polypropylene with PTFE plug.
  • Autoclavable
  • Very good chemical resistance
  • Serrated tubulation on each end accepts 1/2" through 5/16" tubing
Permit reliable, accurate liquid flow control in liquid handling systems. They are ideal for use in developing in-house fluid transfer systems for one-way fluid path for accurate filling or dispensing. The two-way stopcocks feature serrated tubulation on each end.