Urinemonstercontainer, met drukdoppen
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
758901 758902
BRND758901EA 117 EUR
BRND758901 BRND758902
Urinemonstercontainer, met drukdoppen
Diagnostische systemen Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
PP, with red PE lid.
  • Lid is simply sealed by being pressed on
  • Easy to use
  • Simply cut off integrated spout tip and insert test strip
For hygienic and odorless urine sample handling.