Buckets
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
71772 71774
BRND71772EA 18.1 EUR
BRND71772 BRND71774
Buckets
Emmers
HDPE, white.
  • Good chemical and temperature resistance
  • Scale with 1 litre steps
  • Without spout and lid (Please order push-on lid seperately)
For industrial and laboratory applications. With handle.