Methylethylketon ≥99.5% for HPLC
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
EK 500 Additive, Ethyl methyl ketone, 2-Butanone, Butanone, MEK
22924.K2
22924.M1
22924.K2EA
61.5
EUR
22924.K2
22924.M1
Methylethylketon ≥99.5% for HPLC
Methylethylketon
Purity: 99.5+%
CAS No.: 78-93-3
Molecular Formula: C4H8O
Formula Weight: 72.11
Flash Point: -3°(26°F)
Physical Form: Liquid
Density: 0.805
Refractive Index: 1.3790
Appearance: Colourless
Odour: Characteristic
Melting Point: -87°
Boiling Point: 80°
Beilstein Reference: 741880
Merck Reference: 14,6072
UN No.: 1193
MDL No.: MFCD00011648
Formule:
C₂H₅COCH₃ Kookpunt: 80 °C Smeltpunt: –87 °C Dichtheid: 0,805 Vlampunt: –3 °C (26 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011648 CAS nummer: 78-93-3 EINECS: 201-159-0 UN: 1193 ADR: 3,II Bereik: 01-2119457290-43 Merck Index: 14,6072 |