Slang, LDPE, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8010-0125 8010-0250 8010-0375 8010-0500 8010-0625
228-0610EA 52.3 EUR
228-0610 228-0612 228-0613 228-0614 228-0615
Slang, LDPE, Nalgene®
Slangen Transfer Tubing
Made of LDPE, translucent.
  • Contains no plasticizers
  • Nontoxic, odorless, and tasteless
  • Superior chemical resistance
Economically priced and highly suitable for laboratory and industrial use, this low density polyethylene tubing is less flexible than Nalgene 180 PVC tubing but can be bent to facilitate set-ups.
Not autoclavable, but may be chemically sterilized.
Certificaten: Complies with FDA Regulations for food use