Slangen van siliconen
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
143352 143355 143356 143357 143358 143359 143360 143361 143364 143362
BRND143352EA 45.1 EUR
BRND143352 BRND143355 BRND143356 BRND143357 BRND143358 BRND143359 BRND143360 BRND143361 BRND143364 BRND143362
Slangen van siliconen
Slangen Transfer Tubing
Silicone, translucent.
  • Excellent temperature resistance
  • Autoclavable (121 °C)
  • Limited chemical resistance when used with concentrated acids
For laboratory application.