Septumdoppen, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
342178-0384 342178-0240
NALG342178-0384EA 1020 EUR
NALG342178-0384 NALG342178-0240
Septumdoppen, Nalgene®
Sluitingen Flesdoppen
HDPE with silicone/PTFE septum.
  • Sterile 10⁻⁶ SAL
  • No adhesives used to attach septum to high density polyethylene closure
  • Non-cytotoxic and non-pyrogenic
  • Dual septum material for improved gas barrier
Order Now
Specificaties
Environmentally Preferable Lage impact op de fabricage
Milieu certificering NAR Certificate of Retirement