2,4-Pentaandion, EMSURE® for analysis, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
M-Clarity™ quality level = MQ300
1.09600.0100 1.09600.0500
1.09600.0100EA 22.7 EUR
1.09600.0100 1.09600.0500
2,4-Pentaandion, EMSURE® for analysis, Supelco®
2,4-Pentaandion
Formule: CH₃COCH₂COCH₃
Kookpunt: 140,4 °C (1013 hPa)
Smeltpunt: –23 °C
Dichtheid: 0,96945 g/cm³ (20 °C)
Vlampunt: 30 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008787
CAS nummer: 123-54-6
EINECS: 204-634-0
UN: 2310
ADR: 3 (6.1),
Merck Index: 13,00083
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c85a64bf9c7997e9ab03e421a93e7c71e57b8a3&vid=134ed3029693deef3759692be256c57b3a58b087&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Solubility 200 g/l (20 °C)
Melting Point -23 °C
Molar Mass 100.12 g/mol
Boiling Point 138 - 140 °C (1013 hPa)
Vapor Pressure 9 hPa (20 °C)
Flash Point 34 °C
Explosion Limit 2.4 - 11.4 %(V)
Density 0.97 g/cm3 (20 °C)
pH 6 (200 g/l, H2O, 20 °C)
Ignition Point 340 °C
Evaporation number 20