Isobutylacetaat ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
2-Methylpropyl acetate
A14892.0F
A14892.AK
A14892.AP
A14892.0FEA
73.5
EUR
A14892.0F
A14892.AK
A14892.AP
Isobutylacetaat ≥98%
Isobutylacetaat
Purity: 98%
CAS No.: 110-19-0
Molecular Formula: CH3CO2CH2CH(CH3)2
Formula Weight: 116.16
Flash Point: 18°(64°F)
Physical Form: Liquid
Density: 0.870
Refractive Index: 1.3895
Odour: Fruit-like
Melting Point: -99°
Boiling Point: 116-117°
Beilstein Reference: 1741909
Merck Reference: 14,5130
UN No.: 1213
MDL No.: MFCD00008932
Formule:
C₆H₁₂O₂ MW: 116.16 g/mol Kookpunt: 116…117 °C Smeltpunt: –99 °C Dichtheid: 0.870 Vlampunt: 18 °C (64 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008932 CAS nummer: 110-19-0 EINECS: 203-745-1 UN: 1213 ADR: 3 II Merck Index: 13,05147 |