Lauric acid ≥98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Dodecanoic acid, Dodecylic acid
A11672.0C A11672.0E A11672.36
A11672.0CEA 277 EUR
A11672.0C A11672.0E A11672.36
Lauric acid ≥98%
Lauric acid
Purity: 98% CAS No.: 143-07-7 Molecular Formula: CH3(CH2)10CO2H Formula Weight: 200.32 Flash Point: 165°(329°F) Odour: Odourless Boiling Point: 180°/21mm Beilstein Reference: 1099477 Merck Reference: 14,5384 MDL No.: MFCD00002736
Formule: C₁₂H₂₄O₂
MW: 200.32 g/mol
Kookpunt: 180 °C (21 mmHg)
Smeltpunt: 43…45 °C
Dichtheid: 0.883
Vlampunt: 165 °C (329 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002736
CAS nummer: 143-07-7
EINECS: 205-582-1
Merck Index: 13,05400