Lauric acid zS, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Dodecanoic acid, Dodecylic acid
MQ200
8.05333.0100 8.05333.1000 8.05333.9050 8.05333.9010
8.05333.0100EA 32.3 EUR
8.05333.0100 8.05333.1000 8.05333.9050 8.05333.9010
Lauric acid zS, Sigma-Aldrich®
Lauric acid
Formule: C₁₂H₂₄O₂
MW: 200.32 g/mol
Kookpunt: 296
Smeltpunt: 42…45 °C
Dichtheid: 0.875 g/cm³ (20 °C)
Vlampunt: >150 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002736
CAS nummer: 143-07-7
EINECS: 205-582-1
Merck Index: 13,05400
Order Now
Specification Test Results
Solubility 0.058 g/l (20 °C)
Melting Point 43 - 45 °C
Molar Mass 200.32 g/mol
Bulk Density 490 kg/m3
Boiling Point 299 °C (1013 hPa)
Vapor Pressure 0.0027 Pa (25 °C)
Flash Point 176 °C
Explosion Limit 0.6 %(V)
Density 1.03 g/cm3 (20 °C)
Ignition Point 250 °C