Potassium acetate, anhydrous 99+%, pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Acetic acid potassium salt
220150010 220151000 220150050 220150250
ACRO220150010EA 60.3 EUR
ACRO220150010 ACRO220151000 ACRO220150050 ACRO220150250
Potassium acetate, anhydrous 99+%, pure
Potassium acetate
Formule: CH₃COOK
MW: 98.14 g/mol
Smeltpunt: 292 °C
Vlampunt: >250 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012458
CAS nummer: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White Crystalline or granular powder
Infrared spectrum Conforms
Titration with HClO4 ≥99.0 %
Specificaties
Environmentally Preferable