Barium acetate ≥99%, Puriss. p.a. ACS, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

32305-50KG 32305-1KG
32305-50KGEA 1880 EUR
32305-50KG 32305-1KG
Barium acetate ≥99%, Puriss. p.a. ACS, Fluka™
Barium acetate
Formule: (H₃CCOO)₂Ba
Kookpunt: Dec. (on heating)
Smeltpunt: ∼450 °C
Dichtheid: 2,46 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012447
CAS nummer: 543-80-6
EINECS: 208-849-0
ADR: 6.1,III
Merck Index: 13,00968

Order Now

Specification Test Results

Assay 99.0 - 102.0 %
Water insoluble matter Max. 0.005 %
pH (5 %, 20°C) 7 - 8.5
Calcium (Ca) Max. 0.002 %
Iron (Fe) Max. 0.0005 %
Potassium (K) Max. 0.003 %
Magnesium (Mg) Max. 0.001 %
Sodium (Na) Max. 0.005 %
Strontium (Sr) Max. 0.2 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 0.0005 %
Oxidizing substances (as NO₃) Max. 0.005 %
KMnO₄ reducing matter (as HCOOH) Max. 0.025 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR