1-Chlorododecane 99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
257102500
257100010
ACRO257102500EA
29.2
EUR
ACRO257102500
ACRO257100010
1-Chlorododecane 99%
1-Chlorododecane
Formule:
C₁₂H₂₅Cl Kookpunt: 260 °C (760 mmHg) Dichtheid: 0,87 g/cm³ Vlampunt: 130 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00013688 CAS nummer: 112-52-7 EINECS: 203-981-5 UN: 3082 ADR: 9,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4420 to 1.4440 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...