Methyleenchloride Stabilised, EMPROVE® ESSENTIAL NF, BP, extra_pure suitable for use as excipient, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Methylene chloride
Stabilisatie: 50 ppm 2-Methyl-2-buten
1.06049.2500 1.06049.6010 1.06049.6025 1.06049.6190
1.06049.2500EA 244 EUR
1.06049.2500 1.06049.6010 1.06049.6025 1.06049.6190
Methyleenchloride Stabilised, EMPROVE® ESSENTIAL NF, BP, extra_pure suitable for use as excipient, SAFC®
Methyleenchloride
Formule: CH₂Cl₂
MW: 84.93 g/mol
Kookpunt: 39.8
Smeltpunt: –95 °C
Dichtheid: 1.322 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000881
CAS nummer: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM