Chloroacetaldehyde dimethyl acetal 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1-Chloro-2,2-dimethoxyethane
108500010 108502500
ACRO108500010EA 112 EUR
ACRO108500010 ACRO108502500
Chloroacetaldehyde dimethyl acetal 97%
Chloroacetaldehyde dimethyl acetal
Formule: C₄H₉ClO₂
Kookpunt: 128 °C (1013 hPa)
Smeltpunt: –73 °C
Dichtheid: 1,09 g/cm³
Vlampunt: 28 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000948
CAS nummer: 97-97-2
EINECS: 202-624-0
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4140 to 1.4160 (20°C, 589 nm)
GC ≥96.0 %