4-Chloro-1-butanol tech. 85%, Technical Grade
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
L04042.18 L04042.30
L04042.18EA 69.2 EUR
L04042.18 L04042.30
4-Chloro-1-butanol tech. 85%, Technical Grade
4-Chloro-1-butanol
Purity: 85% CAS No.: 928-51-8 Molecular Formula: C4H9ClO Formula Weight: 108.57 Flash Point: 66°(151°F) Refractive Index: 1.4570 Boiling Point: 50-52°/1mm Beilstein Reference: 1731408 MDL No.: MFCD00002967
Formule: C₄H₉ClO
Kookpunt: 50…52 °C (1 mmHg)
Dichtheid: 1,089
Vlampunt: 36 °C (97 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002967
CAS nummer: 928-51-8
EINECS: 213-175-5
UN: 2924
ADR: 3,III