Hexyl isocyanate 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

411660250 411661000 411660050
ACRO411660250EA 158 EUR
ACRO411660250 ACRO411661000 ACRO411660050
Hexyl isocyanate 99%
Hexyl isocyanate
Formule: C₇H₁₃NO
Kookpunt: 44 °C (1013 hPa)
Dichtheid: 0,873 g/cm³
Vlampunt: 59 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00037047
CAS nummer: 2525-62-4
EINECS: 219-763-8
UN: 2478
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid (May develop some turbidity or precipitate)
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4220 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR