Octyl isocyanate 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
416460100 416460050
ACRO416460100EA 131 EUR
ACRO416460100 ACRO416460050
Octyl isocyanate 99%
Octyl isocyanate
Formule: C₉H₁₇NO
Kookpunt: 78 °C (1013 hPa)
Dichtheid: 0,88 g/cm³ (20 °C)
Vlampunt: 71 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013887
CAS nummer: 3158-26-7
EINECS: 221-598-1
UN: 2206
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear to hazy colorless to white to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4290 to 1.4320 (20°C, 589 nm)
GC ≥98.5 %
Titration with HClO4 ≥97.5 %