Pentyl isocyanate 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 1-Pentyl isocyanate
314320050 314320010
ACRO314320050EA 107 EUR
ACRO314320050 ACRO314320010
Pentyl isocyanate 98%
Pentyl isocyanate
Formule: C₆H₁₁NO
Kookpunt: 136…137 °C
Dichtheid: 0,87 g/cm³
Vlampunt: 32 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00144846
CAS nummer: 3954-13-0
UN: 3080
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4130 to 1.4150 (20°C, 589 nm)
GC ≥97.5 %