Propylamine for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 1-Propylamine, 1-Aminopropane
M-Clarity™ quality level = MQ200
8.07477.0100 8.07477.0500 8.07477.1000 8.07477.9040
8.07477.0100EA 33.7 EUR
8.07477.0100 8.07477.0500 8.07477.1000 8.07477.9040
Propylamine for synthesis, Sigma-Aldrich®
Propylamine
Formule: H₃CCH₂CH₂NH₂
Kookpunt: 48 °C (1013 hPa)
Smeltpunt: –83 °C
Dichtheid: 0,7175 g/cm³ (20 °C)
Vlampunt: –30 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008205
CAS nummer: 107-10-8
EINECS: 203-462-3
UN: 1277
ADR: 3 (8),II
Merck Index: 12,08028
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point -83 °C
Molar Mass 59.11 g/mol
Boiling Point 48 - 50 °C
Vapor Pressure 330 hPa (20 °C)
Flash Point -30 °C
Refractive Index 1.389 (20 °C, 589 nm)
Explosion Limit 2.1 - 13.6 %(V)
Density 0.72 g/cm3 (20 °C)
pH 12.6 (100 g/l, H2O, 20 °C)
Ignition Point 325 °C