1-Butaanol ≥99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
L13171.AE
L13171.AU
L13171.0F
L13171.AEEA
25.6
EUR
L13171.AE
L13171.AU
L13171.0F
1-Butaanol ≥99%
1-Butaanol
Purity: 99%
CAS No.: 71-36-3
Molecular Formula: CH3(CH2)3OH
Formula Weight: 74.12
Flash Point: 35°(95°F)
Refractive Index: 1.3990
Odour: Alcohol-like
pH: 7
Melting Point: -90 to -89°
Boiling Point: 117-118°
Beilstein Reference: 969148
Merck Reference: 14,1540
UN No.: 1120
MDL No.: MFCD00002964
Formule:
H₃C(CH₂)₂CH₂OH Kookpunt: 117…118 °C Smeltpunt: –90…–89 °C Dichtheid: 0,810 Vlampunt: 35 °C (95 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002964 CAS nummer: 71-36-3 EINECS: 200-751-6 UN: 1120 ADR: 3,III Bereik: 01-2119484630-38 Merck Index: 14,1540 |