2-Propanol, dry_extra over Molecular Sieve 99.5%, AcroSeal®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
364401000 364400010 364400025 364405000
364401000.EA 24.2 EUR
364401000. 364400010. 364400025. 364405000.
2-Propanol, dry_extra over Molecular Sieve 99.5%, AcroSeal®
2-Propanol
Formule: (CH₃)₂CHOH
MW: 60.1 g/mol
Kookpunt: 81…83 °C (760 mmHg)
Smeltpunt: –89.5 °C
Dichtheid: 0.785 g/cm³
Vlampunt: 12 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011674
CAS nummer: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
Bereik: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤0.005 % (Coulometric)
GC ≥99.5 %