2-Propanol ≥99.7% for gas chromatography
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
Purity: 99.7+%
CAS No.: 67-63-0
Molecular Formula: (CH3)2CHOH
Formula Weight: 60.10
Flash Point: 12°(53°F)
Physical Form: Liquid
Density: 0.785
Refractive Index: 1.3770
Odour: Alcohol-like
Melting Point: -90°
Boiling Point: 82-83°
Beilstein Reference: 635639
Merck Reference: 14,5208
UN No.: 1219
MDL No.: MFCD00011674
Suitable for environmental, pesticide, and GC analysis
Formule:
(CH₃)₂CHOH Kookpunt: 82…83 °C Smeltpunt: –90 °C Dichtheid: 0,785 Vlampunt: 12 °C (53 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011674 CAS nummer: 67-63-0 EINECS: 200-661-7 UN: 1219 ADR: 3,II Bereik: 01-2119457558-25 Merck Index: 14,5208 |