3-Pentanol for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
M-Clarity™ quality level = MQ200
8.07502.0100 8.07502.0500
8.07502.0100EA 67.1 EUR
8.07502.0100 8.07502.0500
3-Pentanol for synthesis, Sigma-Aldrich®
3-Pentanol
Formule: C₅H₁₂O
Kookpunt: 116 °C (1013 hPa)
Smeltpunt: –8 °C
Dichtheid: 0,82 g/cm³ (20 °C)
Vlampunt: 33 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004574
CAS nummer: 584-02-1
EINECS: 209-526-7
UN: 1105
ADR: 3,III
Merck Index: 13,07197
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ec7192926b50615dfac930445c63174c6221a44&vid=a2f1fa3f1ffa1ecbfb6a5dd1d0f29da27d1d8737&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Solubility 55 g/l (30 °C)
Melting Point -8 °C
Molar Mass 88.14 g/mol
Boiling Point 115 - 116 °C (1013 hPa)
Vapor Pressure 11.69 hPa (25 °C)
Flash Point 33 °C
Refractive Index 1.4096 (20 °C, 589 nm)
Explosion Limit 1.2 - 8 %(V)
Density 0.82 g/cm3 (20 °C)
Ignition Point 360 °C
Hazard Class 3
Hazard Class Description Flammable Liquid