1,6-Hexanediol ≥97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Hexamethylene glycol, 1,6-Dihydroxy hexane
A12439.0E
A12439.30
A12439.36
A12439.0EEA
152
EUR
A12439.0E
A12439.30
A12439.36
1,6-Hexanediol ≥97%
1,6-Hexanediol
Purity: 97%
CAS No.: 629-11-8
Molecular Formula: HO(CH2)6OH
Formula Weight: 118.18
Flash Point: 147°(296°F)
Physical Form: Cryst solid
Density: 1.120
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 40-43°
Boiling Point: 248-250°
Beilstein Reference: 1633461
Merck Reference: 14,4690
MDL No.: MFCD00002985
Formule:
C₆H₁₄O₂ Kookpunt: 248…250 °C Smeltpunt: 40…43 °C Dichtheid: 1,120 Vlampunt: 147 °C (296 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002985 CAS nummer: 629-11-8 EINECS: 211-074-0 Merck Index: 14,4690 |