Glycerine, Ultrapure for spectrophotometry
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
32450.AE
32450.K2
32450.K7
32450.M1
32450.AEEA
46.7
EUR
32450.AE
32450.K2
32450.K7
32450.M1
Glycerine, Ultrapure for spectrophotometry
Glycerine
CAS No.: 56-81-5
Molecular Formula: HOCH2CH(OH)CH2OH
Formula Weight: 92.09
Flash Point: 160°(320°F)
Physical Form: Viscous liquid
Density: 1.26
Refractive Index: 1.474
Sensitivity: Hygroscopic
Appearance: Colourless
Odour: Odourless
Melting Point: 18°
Boiling Point: 182°/20mm
Beilstein Reference: 635685
Merck Reference: 14,4484
MDL No.: MFCD00004722
Formule:
HOCH₂CH(OH)CH₂OH MW: 92.09 g/mol Kookpunt: 182 °C (20 mmHg) Smeltpunt: 18 °C Dichtheid: 1.26 Vlampunt: 160 °C (320 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004722 CAS nummer: 56-81-5 EINECS: 200-289-5 Merck Index: 14,4484 |