1,2,6-Hexanetriol for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
8.41808.0050 8.41808.9010
8.41808.0050EA 113 EUR
8.41808.0050 8.41808.9010
1,2,6-Hexanetriol for synthesis, Sigma-Aldrich®
1,2,6-Hexanetriol
Formule: C₆H₁₄O₃
Kookpunt: 323 °C (1013 hPa)
Smeltpunt: 25…32 °C
Dichtheid: 1,11 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002976
CAS nummer: 106-69-4
EINECS: 203-424-6
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point 25 - 32 °C
Molar Mass 134.17 g/mol
Boiling Point 178 °C (6.5 hPa)
Vapor Pressure 0.01 hPa (20 °C)
Flash Point 195 °C
Density 1.11 g/cm3 (20 °C)
pH 4.5 - 7.5 (300 g/l, H2O, 25 °C)