1-Heptanethiol ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

L07079.14 L07079.22
L07079.14EA 52.1 EUR
L07079.14 L07079.22
1-Heptanethiol ≥98%
1-Heptanethiol

Purity: 98% CAS No.: 1639-09-4 Molecular Formula: CH3(CH2)6SH Formula Weight: 132.27 Flash Point: 46°(114°F) Refractive Index: 1.4515 Melting Point: -43° Boiling Point: 173-175° Beilstein Reference: 1731687 UN No.: 3336 MDL No.: MFCD00004911

Formule: C₇H₁₆S
Kookpunt: 173…175 °C
Smeltpunt: –43 °C
Dichtheid: 0,854
Vlampunt: 46 °C (114 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004911
CAS nummer: 1639-09-4
EINECS: 216-678-8
UN: 3336
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR