1-Hexadecanethiol, (max. 4% H₂O) 97% (dry weight)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
n-Hexadecyl mercaptan, 1-Mercaptohexadecane, Hexadecane-1-thiol, n-Hexadecanethiol
L15099.AC
L15099.AE
L15099.AP
L15099.ACEA
26.9
EUR
L15099.AC
L15099.AE
L15099.AP
1-Hexadecanethiol, (max. 4% H₂O) 97% (dry weight)
1-Hexadecanethiol
Purity: 97%
CAS No.: 2917-26-2
Molecular Formula: C16H34S
Formula Weight: 258.51
Flash Point: 135°(275°F)
Refractive Index: 1.4625
Odour: Unpleasant
Melting Point: 19-23°
Boiling Point: 334°
Beilstein Reference: 1748495
MDL No.: MFCD00011677
Formule:
C₁₆H₃₄S Kookpunt: 334 °C Smeltpunt: 19…23 °C Dichtheid: 0,846 Vlampunt: 135 °C (275 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011677 CAS nummer: 2917-26-2 EINECS: 220-846-6 UN: 3334 ADR: 9, |