1-Octanethiol 97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
157480010
157482500
157480025
157480020
ACRO157480010EA
93.7
EUR
ACRO157480010
ACRO157482500
ACRO157480025
ACRO157480020
1-Octanethiol 97%
1-Octanethiol
Formule:
C₈H₁₈S Kookpunt: 197…200 °C (760 mmHg) Smeltpunt: –49 °C Dichtheid: 0,84 g/cm³ Vlampunt: 68 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004912 CAS nummer: 111-88-6 EINECS: 203-918-1 UN: 3082 ADR: 9,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4525 to 1.4545 (20°C, 589 nm) |
GC | ≥96.0 % |
Titration with NaOH | ≥96.0 to ≤104.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...