1-Octanethiol for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
n-Octyl mercaptan, 1-Mercaptooctane, n-Octanethiol
M-Clarity™ quality level = MQ200
8.21861.0100
8.21861.0500
8.21861.0100EA
38.7
EUR
8.21861.0100
8.21861.0500
1-Octanethiol for synthesis, Sigma-Aldrich®
1-Octanethiol
Formule:
C₈H₁₈S Kookpunt: 199 °C (1013 hPa) Smeltpunt: –49 °C Dichtheid: 0,84 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004912 CAS nummer: 111-88-6 EINECS: 203-918-1 UN: 3082 ADR: 9,III |
Specification Test Results
Solubility | 0.004 g/l (20 °C) |
Melting Point | -49 °C |
Molar Mass | 146.29 g/mol |
Boiling Point | 199 °C (1013 hPa) |
Vapor Pressure | 0.3 hPa (20 °C) |
Flash Point | 70 °C |
Refractive Index | 1.4538 (20 °C, 589 nm) |
Density | 0.84 g/cm3 (20 °C) |
Ignition Point | 240 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...