n-Heptaan 99+%, extra pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: high purity solvents
383790025 383790050 383790010 383790250
ACRO383790025EA 128 EUR
ACRO383790025 ACRO383790050 ACRO383790010 ACRO383790250
n-Heptaan 99+%, extra pure
n-Heptaan
Formule: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Kookpunt: 98 °C (760 mmHg)
Smeltpunt: –91 °C
Dichtheid: 0.683 g/cm³
Vlampunt: –4 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009544
CAS nummer: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3 II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥99.0 %
Residue after evaporation ≤20 ppm