n-Heptaan, anhydrous 99+%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
high purity solvents
Packaged under argon in resealable ChemSeal™ bottles.
44457.AE
44457.K2
44457.AEEA
22.9
EUR
44457.AE
44457.K2
n-Heptaan, anhydrous 99+%
n-Heptaan
Purity: 99+%
CAS No.: 142-82-5
Molecular Formula: C7H16
Formula Weight: 100.21
Flash Point: -4°(24°F)
Physical Form: Liquid
Density: 0.684
Refractive Index: 1.3877
Appearance: Colourless
Odour: Petroleum-like
Melting Point: -91°
Boiling Point: 97-99°
Beilstein Reference: 1730763
Merck Reference: 14,4659
UN No.: 1206
MDL No.: MFCD00009544
Formule:
H₃C(CH₂)₅CH₃ MW: 100.20 g/mol Kookpunt: 97…99 °C Smeltpunt: –91 °C Dichtheid: 0.684 Vlampunt: –4 °C (24 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00009544 CAS nummer: 142-82-5 EINECS: 205-563-8 UN: 1206 ADR: 3 II Merck Index: 14,4659 |