Hexane (mixture of isomers) ≥98.5% ACS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
33321.K2 33321.M1 33321.K4
33321.K2EA 44.9 EUR
33321.K2 33321.M1 33321.K4
Hexane (mixture of isomers) ≥98.5% ACS
Hexane (mixture of isomers)
Purity: 98.5+% CAS No.: 92112-69-1 Molecular Formula: C6H14 Formula Weight: 86.18 Flash Point: -23°(-9°F) Physical Form: Liquid, Mixture of 5 Isomers Density: 0.67 Refractive Index: 1.3795 Appearance: Colourless Melting Point: -95° Boiling Point: 65-70° UN No.: 1208 MDL No.: MFCD02179311
Formule: H₃C(CH₂)₄CH₃
Kookpunt: 65…70 °C
Smeltpunt: –95 °C
Dichtheid: 0,67
Vlampunt: <–23 °C (–9 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02179311
CAS nummer: 110-54-3
EINECS: 295-570-2
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM