n-Hexaan 95 HPLC
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Hexyl hydride, Hexanen, Hexane
39199.K2
39199.M1
39199.K4
39199.K2EA
79.1
EUR
39199.K2
39199.M1
39199.K4
n-Hexaan 95 HPLC
n-Hexaan
Purity: 95% min
CAS No.: 110-54-3
Molecular Formula: CH3(CH2)4CH3
Formula Weight: 86.18
Flash Point: -23°(-9°F)
Physical Form: Liquid, Distilled in Glass
Density: 0.659
Refractive Index: 1.3750
Odour: Weak, characteristic
Melting Point: -95°
Boiling Point: 69°
Beilstein Reference: 1730733
Merck Reference: 14,4694
UN No.: 1208
MDL No.: MFCD00009520
Formule:
H₃C(CH₂)₄CH₃ MW: 86.18 g/mol Kookpunt: 69 °C Smeltpunt: –95 °C Dichtheid: 0.659 Vlampunt: –23 °C (–9 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00009520 CAS nummer: 110-54-3 EINECS: 203-777-6 UN: 1208 ADR: 3 II Merck Index: 14,4694 |