Qualitative filter papers, technical, creped
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FT-2-483580580 FT-2-342580580 FT-4-423-125 FT-3-423-090 FT2-335-580580 FT-3-423-240 FT-3-335-125 FT-3-423-185 FT-3-423-150 FT-3-335-270 FT-3-335-150 FT-3-335-110 FT-3-335-185 FT-4-423-185 FT-4-423-385 FT-4-423-320 FT-4-423-240 FT-4-423-150 FT-4-423-270 FT-3-314-150 FT-3-314-090 FT-4-423-110 FT-2-354580580 FT-2-423580580 FT-4-483-185 FT-4-480-150 FT-2-314580580 FT-4-483-500 FT-3-342-125 FT-3-354-185 FT-3-354-240 FT-3-354-320 FT-3-423-110 FT-3-423-125 FT-4-483-240 FT-3-480-185 FT-3-314-110 FT-3-480-125 FT-3-335-090 FT-3-335-190 FT-3-335-200 FT-3-480-150 FT-3-314-070 FT-3-314-240 FT-3-314-385 FT-4-335-125 FT-4-335-240 FT-4-335-320 FT-4-423-500 FT2-480-580580
515-1613EA 489 EUR
515-1613 515-1623 SARTFT-4-423-125 SARTFT-3-423-090 512-0536 SARTFT-3-423-240 SARTFT-3-335-125 SARTFT-3-423-185 SARTFT-3-423-150 SARTFT-3-335-270 SARTFT-3-335-150 SARTFT-3-335-110 SARTFT-3-335-185 SARTFT-4-423-185 SARTFT-4-423-385 SARTFT-4-423-320 SARTFT-4-423-240 SARTFT-4-423-150 SARTFT-4-423-270 SARTFT-3-314-150 SARTFT-3-314-090 SARTFT-4-423-110 SARTFT-2-354580580 SARTFT-2-423580580 SARTFT-4-483-185 SARTFT-4-480-150 SARTFT-2-314580580 SARTFT-4-483-500 SARTFT-3-342-125 SARTFT-3-354-185 SARTFT-3-354-240 SARTFT-3-354-320 SARTFT-3-423-110 SARTFT-3-423-125 SARTFT-4-483-240 SARTFT-3-480-185 SARTFT-3-314-110 SARTFT-3-480-125 SARTFT-3-335-090 SARTFT-3-335-190 SARTFT-3-335-200 SARTFT-3-480-150 SARTFT-3-314-070 SARTFT-3-314-240 SARTFT-3-314-385 SARTFT-4-335-125 SARTFT-4-335-240 SARTFT-4-335-320 SARTFT-4-423-500 SARTFT2-480-580580
Qualitative filter papers, technical, creped
Filters Filterpapier Kwalitatief filterpapier
These filter papers are used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications. Grades with a wet burst resistance >30 kPa are referred to as wet-strengthened and are therefore
suitable for pressure or vacuum filtration.
They are made of refined pulp and linters with an >95% alpha-cellulose content, are very pure with an ash content between <0,1 to 0,15%.
Grade 5 H/N: Very fast rate of filtration, wide-pore. Used for filtration of essential oils.
Grade 37/N: Very fast rate of filtration, wide-pore.
Grade FT 55: Very fast rate of filtration. Used for rapid clarification of coarse contaminants or impurities; galvanic baths; for use as articulation paper for dental impression trays.
Grade 1602/N: Very fast rate of filtration. All-purpose filter papers for even faster clarification than with grade 603 before performing less critical analyses of highly viscous solutions.
Grade 39/N: Very fast rate of filtration, wide-pore. Used for cooking oils and vinegar filtration.
Grade 603/N: Fast rate of filtration. Used for sugar juices; sugar solutions; clarifying filtration of clarified dried sugar beet shred and of beet sugar extracts.
Grade 6 S/N: Medium fast rate of filtration. Used as prefilters for transformer oil.
Grade 601/N: Medium fast rate of filtration.
Grade 17/N: Medium fast rate of filtration. Used for fine filtration of tar.