Compact burettes, interchangeable PTFE stopcock, SILBERBRAND
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13906 13903 13908
BRND13906EA 68.4 EUR
BRND13906 BRND13903 BRND13908
Compact burettes, interchangeable PTFE stopcock, SILBERBRAND
Buretten
AR-GLAS®.
  • Calibrated to deliver (TD, Ex)
  • Schellbach stripe
  • Reduced distance between subdivision marks
Error limits according to class B.
Certificaten: DIN EN ISO 385
Leveringsinformatie: Supplied as burette tube, stopcock with precision tip.